Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map1lc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007978.2 | NP_001007979.1 | 2 - 3 | 154 | 71 | |
GenBank mRNA | BC080488.1 | - | 2 - 3 | 156 | 71 | |
CR761386.2 | - | 2 - 3 | 200 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |