Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map1lc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007978.2 | NP_001007979.1 | 3 - 4 | 265 | 101 | |
GenBank mRNA | BC080488.1 | - | 3 - 4 | 267 | 101 | |
CR761386.2 | - | 3 - 4 | 311 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |