Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psma4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007997.1 | NP_001007998.1 | 7 - 8 | 641 | 84 | |
XM_018093016.1 | XP_017948505.1 | 7 - 8 | 695 | 84 | ||
XM_018093017.1 | XP_017948506.1 | 7 - 8 | 727 | 84 | ||
GenBank mRNA | BC080876.1 | - | 7 - 8 | 641 | 84 | |
CR761624.2 | - | 7 - 8 | 616 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |