Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdf2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008032.1 | NP_001008033.1 | 2 - 3 | 400 | 72 | |
GenBank mRNA | BC080914.1 | - | 2 - 3 | 400 | 72 | |
CR761046.2 | - | 2 - 3 | 378 | 72 |
Gene Symbol: LOC101732749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004920857.2 | XP_004920914.1 | 2 - 3 | 622 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |