Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alkbh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008037.1 | NP_001008038.1 | 3 - 4 | 1012 | 72 | |
GenBank mRNA | BC080920.1 | - | 3 - 4 | 1012 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |