Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ears2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008041.1 | NP_001008042.1 | 3 - 4 | 506 | 86 | |
GenBank mRNA | BC080925.1 | - | 3 - 4 | 506 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |