Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008159.1 | NP_001008160.1 | 3 - 4 | 261 | 91 | |
GenBank mRNA | BC082340.1 | - | 3 - 4 | 261 | 91 | |
CR760752.1 | - | 3 - 4 | 254 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |