Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008160.1 | NP_001008161.1 | 2 - 3 | 337 | 103 | |
XM_012960964.1 | XP_012816418.1 | 1 - 2 | 276 | 103 | ||
GenBank mRNA | BC082341.1 | - | 2 - 3 | 337 | 103 | |
CR761171.2 | - | 1 - 2 | 293 | 103 | ||
CR762107.2 | - | 1 - 2 | 122 | 103 | ||
CU075362.1 | - | 1 - 2 | 282 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |