Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008445.1 | NP_001008445.1 | 5 - 6 | 731 | 105 | |
GenBank mRNA | BC080453.1 | - | 5 - 6 | 731 | 105 | |
CR855667.2 | - | 5 - 6 | 643 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |