Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008445.1 | NP_001008445.1 | 6 - 7 | 824 | 104 | |
GenBank mRNA | BC080453.1 | - | 6 - 7 | 824 | 104 | |
CR855667.2 | - | 6 - 7 | 736 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |