Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011096.1 | NP_001011096.1 | 4 - 5 | 534 | 113 | |
GenBank mRNA | BC084490.1 | - | 4 - 5 | 534 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |