Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011116.1 | NP_001011116.1 | 2 - 3 | 398 | 79 | |
GenBank mRNA | BC084514.1 | - | 2 - 3 | 398 | 79 | |
CR760847.2 | - | 2 - 3 | 373 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |