Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011170.1 | NP_001011170.1 | 5 - 6 | 609 | 74 | |
XM_012960989.2 | XP_012816443.1 | 5 - 6 | 590 | 74 | ||
GenBank mRNA | BC084996.1 | - | 5 - 6 | 609 | 74 | |
CR760886.2 | - | 5 - 6 | 542 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |