Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011257.1 | NP_001011257.1 | 10 - 11 | 778 | 79 | |
XM_012968299.2 | XP_012823753.1 | 9 - 10 | 800 | 79 | ||
GenBank mRNA | BC087567.2 | - | 10 - 11 | 778 | 79 | |
CR855724.2 | - | 10 - 11 | 749 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |