Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011270.1 | NP_001011270.1 | 8 - 9 | 1101 | 76 | |
XM_012958933.2 | XP_012814387.1 | 9 - 10 | 1507 | 76 | ||
XM_012958934.2 | XP_012814388.1 | 8 - 9 | 1314 | 76 | ||
XM_012958935.2 | XP_012814389.1 | 8 - 9 | 2041 | 76 | ||
XM_012958937.2 | XP_012814391.1 | 7 - 8 | 958 | 76 | ||
XM_018092056.1 | XP_017947545.1 | 9 - 10 | 1503 | 76 | ||
GenBank mRNA | BC087979.1 | - | 8 - 9 | 1101 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |