Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203571.1 | NP_988902.1 | 4 - 5 | 409 | 83 | |
XM_012956621.2 | XP_012812075.1 | 4 - 5 | 496 | 83 | ||
GenBank mRNA | BC061625.1 | - | 4 - 5 | 409 | 83 | |
CR760224.2 | - | 4 - 5 | 414 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |