Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203577.1 | NP_988908.1 | 3 - 4 | 288 | 100 | |
XM_012966262.1 | XP_012821716.1 | 3 - 4 | 324 | 100 | ||
XM_012966263.2 | XP_012821717.1 | 3 - 4 | 296 | 100 | ||
GenBank mRNA | BC061617.1 | - | 3 - 4 | 288 | 100 | |
BC077651.1 | - | 3 - 4 | 260 | 100 | ||
CR762071.2 | - | 2 - 3 | 224 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |