Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203595.1 | NP_988926.1 | 1 - 2 | 323 | 79 | |
GenBank mRNA | BC061606.1 | - | 1 - 2 | 323 | 79 | |
CR762300.2 | - | 1 - 2 | 205 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |