Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011418.1 | NP_001011418.1 | 4 - 5 | 807 | 63 | |
GenBank mRNA | BC088010.1 | - | 4 - 5 | 807 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |