Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011474.1 | NP_001011474.1 | 3 - 4 | 432 | 104 | |
XM_012961449.2 | XP_012816903.1 | 3 - 4 | 2155 | 104 | ||
XM_012961451.1 | XP_012816905.1 | 4 - 5 | 624 | 104 | ||
GenBank mRNA | BC088764.1 | - | 3 - 4 | 432 | 104 | |
CR848255.2 | - | 3 - 4 | 430 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |