Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vsir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011488.1 | NP_001011488.1 | 6 - 7 | 906 | 87 | |
GenBank mRNA | BC088869.1 | - | 6 - 7 | 906 | 87 |
Gene Symbol: LOC101733739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018096050.1 | XP_017951539.1 | 2 - 3 | 265 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |