Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c2orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015782.1 | NP_001015782.2 | 2 - 3 | 268 | 75 | |
GenBank mRNA | BC089724.1 | - | 2 - 3 | 268 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |