Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015834.1 | NP_001015834.1 | 6 - 7 | 1342 | 94 | |
XM_018092983.1 | XP_017948472.1 | 6 - 7 | 988 | 94 | ||
GenBank mRNA | BC090120.1 | - | 6 - 7 | 1342 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |