Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015849.1 | NP_001015849.1 | 2 - 3 | 144 | 78 | |
XM_012961773.1 | XP_012817227.1 | 2 - 3 | 328 | 78 | ||
GenBank mRNA | BC090139.1 | - | 2 - 3 | 144 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |