Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc50a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015865.1 | NP_001015865.1 | 3 - 4 | 585 | 77 | |
GenBank mRNA | BC090379.1 | - | 3 - 4 | 585 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |