Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015871.1 | NP_001015871.1 | 4 - 5 | 406 | 96 | |
GenBank mRNA | BC090805.1 | - | 4 - 5 | 406 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |