Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016317.2 | NP_001016317.1 | 3 - 4 | 454 | 97 | |
GenBank mRNA | BC158155.1 | - | 3 - 4 | 492 | 97 | |
CR761465.2 | - | 3 - 4 | 454 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |