Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: figla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016342.2 | NP_001016342.1 | 2 - 3 | 241 | 94 | |
XM_012959005.2 | XP_012814459.1 | 2 - 3 | 350 | 94 | ||
GenBank mRNA | BC155532.1 | - | 1 - 2 | 214 | 94 | |
CR761381.2 | - | 2 - 3 | 241 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |