Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c15orf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016489.2 | NP_001016489.1 | 11 - 12 | 805 | 82 | |
XM_012968415.2 | XP_012823869.1 | 11 - 12 | 844 | 82 | ||
XM_012968416.2 | XP_012823870.1 | 11 - 12 | 814 | 82 | ||
XM_012968417.2 | XP_012823871.1 | 11 - 12 | 799 | 82 | ||
GenBank mRNA | BC135523.1 | - | 11 - 12 | 817 | 82 | |
CR760834.2 | - | 11 - 12 | 805 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |