Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016492.2 | NP_001016492.1 | 8 - 9 | 784 | 107 | |
GenBank mRNA | BC127348.1 | - | 7 - 8 | 778 | 107 | |
BC135545.1 | - | 7 - 8 | 759 | 107 | ||
CR760826.2 | - | 8 - 9 | 784 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |