Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016493.2 | NP_001016493.1 | 3 - 4 | 480 | 91 | |
XM_018091262.1 | XP_017946751.1 | 3 - 4 | 354 | 91 | ||
GenBank mRNA | CR760825.2 | - | 3 - 4 | 480 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |