Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016645.2 | NP_001016645.1 | 3 - 4 | 206 | 91 | |
GenBank mRNA | BC135754.1 | - | 3 - 4 | 269 | 91 | |
CR760396.2 | - | 3 - 4 | 206 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |