Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anp32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016746.2 | NP_001016746.1 | 2 - 3 | 271 | 79 | |
GenBank mRNA | BC167942.1 | - | 2 - 3 | 542 | 79 | |
CR855828.2 | - | 2 - 3 | 271 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |