Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016811.1 | NP_001016811.1 | 7 - 8 | 1389 | 140 | |
XM_012968444.2 | XP_012823898.1 | 7 - 8 | 1458 | 140 | ||
GenBank mRNA | BC167578.1 | - | 7 - 8 | 1409 | 140 | |
CR855567.1 | - | 7 - 8 | 1389 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |