Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016968.2 | NP_001016968.1 | 4 - 5 | 315 | 62 | |
XM_012961154.2 | XP_012816608.1 | 4 - 5 | 348 | 62 | ||
GenBank mRNA | CR848466.2 | - | 4 - 5 | 315 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |