Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aimp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017050.2 | NP_001017050.2 | 5 - 6 | 673 | 80 | |
XM_012963225.1 | XP_012818679.1 | 6 - 7 | 944 | 80 | ||
GenBank mRNA | BC075609.1 | - | 5 - 6 | 673 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |