Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017113.2 | NP_001017113.1 | 3 - 4 | 654 | 81 | |
XM_018093906.1 | XP_017949395.1 | 3 - 4 | 901 | 81 | ||
GenBank mRNA | CR761203.2 | - | 3 - 4 | 654 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |