Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3gat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030349.1 | NP_001025520.1 | 2 - 3 | 196 | 85 | |
XM_012954327.2 | XP_012809781.1 | 4 - 5 | 580 | 85 | ||
XM_012954328.2 | XP_012809782.1 | 3 - 4 | 398 | 85 | ||
XM_012954329.2 | XP_012809783.1 | 4 - 5 | 568 | 85 | ||
XM_012954330.2 | XP_012809784.1 | 3 - 4 | 386 | 85 | ||
XM_012954331.2 | XP_012809785.1 | 3 - 4 | 378 | 85 | ||
GenBank mRNA | AJ890837.1 | - | 2 - 3 | 196 | 85 | |
BC121200.1 | - | 4 - 5 | 483 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |