Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030359.1 | NP_001025530.1 | 641 | 63 | ||
GenBank mRNA | BC090566.1 | - | 641 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis |