Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030428.1 | NP_001025599.1 | 2 - 3 | 430 | 84 | |
GenBank mRNA | BC091048.1 | - | 2 - 3 | 430 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |