Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030460.2 | NP_001025631.1 | 1 - 2 | 102 | 76 | |
GenBank mRNA | BC091584.1 | - | 1 - 2 | 106 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |