Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030517.1 | NP_001025688.1 | 5 - 6 | 713 | 65 | |
XM_012959135.1 | XP_012814589.1 | 5 - 6 | 780 | 65 | ||
GenBank mRNA | BC094531.1 | - | 5 - 6 | 713 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |