Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xkr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033942.2 | NP_001029114.1 | 4 - 5 | 728 | 68 | |
XM_012964043.2 | XP_012819497.1 | 4 - 5 | 763 | 68 | ||
XM_018089354.1 | XP_017944843.1 | 3 - 4 | 608 | 68 | ||
GenBank mRNA | AY699708.1 | - | 4 - 5 | 731 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |