Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213675.2 | NP_998840.1 | 2 - 3 | 276 | 70 | |
GenBank mRNA | BC067980.2 | - | 2 - 3 | 276 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |