Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011385.2 | NP_001011385.2 | 5 - 6 | 1254 | 88 | |
XM_018092852.1 | XP_017948341.1 | 6 - 7 | 1634 | 88 | ||
XM_018092853.1 | XP_017948342.1 | 6 - 7 | 1532 | 88 | ||
XM_018092854.1 | XP_017948343.1 | 6 - 7 | 1584 | 88 | ||
XM_018092855.1 | XP_017948344.1 | 5 - 6 | 1197 | 88 | ||
GenBank mRNA | BC088568.1 | - | 5 - 6 | 1177 | 88 | |
CR855420.2 | - | 5 - 6 | 1254 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |