Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016157.2 | NP_001016157.1 | 1 - 2 | 107 | 64 | |
GenBank mRNA | CR761936.2 | - | 1 - 2 | 107 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |