Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016356.2 | NP_001016356.1 | 2 - 3 | 254 | 82 | |
GenBank mRNA | BC135477.1 | - | 2 - 3 | 228 | 82 | |
BC157765.1 | - | 2 - 3 | 267 | 82 | ||
CR761308.2 | - | 2 - 3 | 254 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |