Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016376.2 | NP_001016376.1 | 3 - 4 | 100 | 92 | |
XM_012962994.2 | XP_012818448.1 | 3 - 4 | 296 | 92 | ||
XM_012962995.2 | XP_012818449.1 | 3 - 4 | 285 | 92 | ||
GenBank mRNA | BC153684.1 | - | 3 - 4 | 148 | 92 | |
CR761250.2 | - | 3 - 4 | 100 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |