Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016473.2 | NP_001016473.1 | 6 - 7 | 575 | 70 | |
GenBank mRNA | BC157231.1 | - | 7 - 8 | 777 | 70 | |
CR760906.2 | - | 6 - 7 | 575 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |