Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016518.2 | NP_001016518.1 | 2 - 3 | 472 | 78 | |
GenBank mRNA | BC170850.1 | - | 1 - 2 | 306 | 78 | |
BC170852.1 | - | 1 - 2 | 306 | 78 | ||
CR926213.2 | - | 2 - 3 | 472 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |